Läs mer
N-Nitroso Betahistine, TRC
Produktkod. 17964285
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964285 50 mg
17945085 500 mg
17964995 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN86945050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32635-81-7
Molekylformel C8 H11 N3 O
Molekylvikt (g/mol) 165.192
Synonym N-Methyl-N-nitroso-2-pyridineethanamine, N-Methyl-N-(2-(pyridin-2-yl)ethyl)nitrous amide
IUPAC-namn N-methyl-N-[2-(2-pyridyl)ethyl]nitrous amide
LEDER CN(CCc1ccccn1)N=O
Specifikationer
Kemiskt namn eller material N-Nitroso Betahistine
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C8H11N3O/c1-11(10-12)7-5-8-4-2-3-6-9-8/h2-4,6H,5,7H2,1H3
Formel vikt 165.0902
Procent renhet >95%