Läs mer
N,O-Di(2-hydroxyethyl)-2-amino-5-nitrophenol, TRC
Produktkod. 30376621
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376621 500 mg
30371261 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376621 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD451765500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59820-43-8
Molekylformel C10H14N2O5
Molekylvikt (g/mol) 242.23
LEDER OCCNc1ccc(cc1OCCO)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material N,O-Di(2-hydroxyethyl)-2-amino-5-nitrophenol
Kvantitet 500 mg
InChI formel InChI=1S/C10H14N2O5/c13-4-3-11-9-2-1-8(12(15)16)7-10(9)17-6-5-14/h1-2,7,11,13-14H,3-6H2
Formel vikt 242.0903