Läs mer
N-Pivaloyl-L-tyrosine, TRC
Produktkod. 30410584
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410584 25 mg
30427664 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30410584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP55150025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33019-85-1
Molekylformel C14H19NO4
Molekylvikt (g/mol) 265.3
Synonym N-(2,2-Dimethyl-1-oxopropyl)-L-tyrosine
LEDER Cl.O=C1N(C)c2c(OC1)c(cc(Cl)c2)C(=O)NC3C4CCN(CC4)C3
Specifikationer
Kemiskt namn eller material N-Pivaloyl-L-tyrosine
InChI formel InChI=1S/C14H19NO4/c1-14(2,3)13(19)15-11(12(17)18)8-9-4-6-10(16)7-5-9/h4-7,11,16H,8H2,1-3H3,(H,15,19)(H,17,18)/t11-/m0/s1