Läs mer
N-Stearoyl Phytosphingosine, TRC
Produktkod. 30179550
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179550 50 mg
30179102 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179550 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68658550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34354-88-6
Molekylformel C36 H73 N O4
Molekylvikt (g/mol) 583.97
Synonym N-[(1S,2S,3R)-2,3-Dihydroxy-1-(hydroxymethyl)heptadecyl]octadecanamide, C18 3-Hydroxy Dihydroceramide, Ceramide III, Ceramide 3, DS-Ceramide Y3S, D-ribo-1,3,4-Trihydroxy-2-octadecanoylamidooctadecane, 2S,3S,4R-2-Stearoylamide-1,3,4-octadecanetriol;
IUPAC-namn N-[(2S,3S,4R)-1,3,4-trihydroxyoctadecan-2-yl]octadecanamide
LEDER CCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@H](O)[C@H](O)CCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material N-Stearoyl Phytosphingosine
Kvantitet 50 mg
InChI formel InChI=1S/C36H73NO4/c1-3-5-7-9-11-13-15-17-18-19-21-23-25-27-29-31-35(40)37-33(32-38)36(41)34(39)30-28-26-24-22-20-16-14-12-10-8-6-4-2/h33-34,36,38-39,41H,3-32H2,1-2H3,(H,37,40)/t33-,34+,36-/m0/s1
Formel vikt 583.554