Learn More
Thermo Scientific™ Nalgene™ Colored Polypropylene Closures: Bulk Pack
Eliminate spills with these one-piece, colored, polypropylene closures, which fit narrow-mouth 4mL bottles.
Brand: Thermo Scientific™ 362150-5130
Description
Eliminate spills with Thermo Scientific™ Nalgene™ Colored Polypropylene Closures. These one-piece, linerless closures fit Nalgene™ narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Features:- Guaranteed leakproof†when used with Nalgene narrow-mouth bottles
- Reduce chance of cross contamination
- Protect contents from UV light when used with opaque bottles
- Available in Opaque white, amber, green, red and blue for color coding
Ordering Information:
Case of 2,000.
Specifications
Red | |
Polypropylene | |
2000 | |
13-415 | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Closures meet FDA CFR21 177.1520 for food and beverage use, USP Class VI. Meet the requirements of light-resistant containers per USP latest edition when used with opaque bottles. | |
Round | |
Screw Cap | |
0.315 in. |
Nalgene Colored Closures | |
2,000 | |
Bulk | |
Unlined | |
Narrow | |
8 mm | |
Nalgene narrow-mouth 4mL bottles | |
16 mm (Outer) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7448422010 | Certificate of Compliance | 28/05/2025 | 362150-5130 |
7445906010 | Certificate of Compliance | 22/05/2025 | 362150-5130 |
7442473010 | Certificate of Compliance | 29/04/2025 | 362150-5130 |
7452379010 | Certificate of Compliance | 17/04/2025 | 362150-5130 |
7450764010 | Certificate of Compliance | 05/03/2025 | 362150-5130 |
Note: Not for use on hot plates