Learn More
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack, 4mL
BIOPHARMAGREENER_CHOICE
Safely store and ship light-sensitive, small-volume valuable reagents with these opaque amber HDPE diagnostic bottles, for use in test kits and many small-volume applications.
Brand:  Thermo Scientific™ 312004-9125_MX
Product Code. 10222913
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Safely store and ship light-sensitive, small-volume valuable reagents with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. For use in test kits and many small-volume applications, these high density polyethylene bottles feature opaque amber linerless polypropylene closure that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions. The durable, nonpyrogenic and noncytotoxic uniform walls are resistant to splitting or puncturing.
  • Nonsterile
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.
Documents
Schematics & Diagrams
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1416821 Certificate TypeCertificate of Compliance Date06/11/2024 Catalog Number312004-9125_MX,312004-9125
Lot Number1414693 Certificate TypeCertificate of Compliance Date29/09/2024 Catalog Number312004-9125_MX,312004-9125
Lot Number1413843 Certificate TypeCertificate of Compliance Date18/09/2024 Catalog Number312004-9125_MX,312004-9125
Lot Number1398951 Certificate TypeCertificate of Compliance Date20/06/2024 Catalog Number312004-9125
Lot Number1404981 Certificate TypeCertificate of Compliance Date19/05/2024 Catalog Number312004-9125_MX,312004-9125
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack, 4mL > 3.4mL, 20-415 closure, bulk pack
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.