Learn More
Thermo Scientific™ Nalgene™ PETG Certified Clean Containers, 1000mL
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. 
Brand:  Thermo Scientific™ 382019-1000
Product Code. 16470581
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier propertiesand ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1428396 Certificate TypeCertificate of Processing Date11/06/2025 Catalog Number382019-1000
Lot Number1428011 Certificate TypeCertificate of Processing Date16/05/2025 Catalog Number382019-1000
Lot Number1423843 Certificate TypeCertificate of Processing Date29/04/2025 Catalog Number382019-1000
Lot Number1428396 Certificate TypeCertificate of Compliance Date14/05/2025 Catalog Number382019-1000
Lot Number1428011 Certificate TypeCertificate of Compliance Date01/05/2025 Catalog Number382019-1000
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ PETG Certified Clean Containers, 1000mL > 1000mL; 24/Cs.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.