Learn More
Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents.
Brand: Thermo Scientific™ 6320-0010
360.39 SEK valid until 2025-12-31
Use promo code "24083" to get your promotional price.
Description
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents. Double-action hinges provide extra leverage and long life. These forceps feature a scissor-type handle with a ratchet to lock tips shut and a handy clamp for chromatography columns.
- Autoclavable
- Feature jaws with interlocking teeth to grip objects tightly
Specifications
Straight, Serrated | |
Polypropylene | |
Forceps | |
Scissor Type |
12 | |
Forceps | |
12/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7436783010 | Certificate of Compliance | 11/06/2025 | 6320-0010 |
7448424010 | Certificate of Compliance | 30/05/2025 | 6320-0010 |
7452535010 | Certificate of Compliance | 31/03/2025 | 6320-0010 |
7401987010 | Certificate of Compliance | 15/03/2025 | 6320-0010 |
7174064010 | Certificate of Compliance | 15/03/2025 | 6320-0010 |