missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NAPB |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892594
|
Proteintech
14106-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882594
|
Proteintech
14106-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Required for vesicular transport between the endoplasmic reticulum and the Golgi apparatus.Specifications
NAPB | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P28663, P85969, Q9H115 | |
17957, 499903, 63908 | |
NAPB Fusion Protein Ag5239 | |
Primary | |
-20°C | |
NAPB |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
beta SNAP, NAPB, SNAP BETA, SNAPB | |
Napb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NAPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech