missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14882-1-AP-150UL
This item is not returnable.
View return policy
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases.Specifications
NARS | |
Polyclonal | |
Unconjugated | |
NARS | |
ASNRS, Asparagine tRNA ligase, asparaginyl tRNA synthetase, NARS, NARS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291556, 4677, 70223 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43776, Q8BP47 | |
Nars | |
NARS Fusion Protein Ag6656 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction