Learn More
Thermo Scientific™ National Target DP™ Vial Kits
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
545.00 SEK
Product Code | Brand | Marking Spot | Color (Seal) | Material | Septum | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Marking Spot | Color (Seal) | Material | Septum | Price | Quantity & Availability | |||||
11971563
|
Thermo Scientific™
C4000-97 |
No | Red/White | Polypropylene | PTFE/Silicone | N/A | ||||||
Description
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
- Convenience Kits with 2mL, 12 Ă— 32mm vials
- I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
- Blue caps with your choice of vial and preassembled seals with septa
- Packaged in two-compartment benchtop storage tray
- All items remain dust-free
- Caps with pre-inserted septa are not attached to vials (packed in separate polybag)
Specifications
Blue | |
9-425 |