Learn More
Ark Plas Products™ Natural Polypropylene Commercial Grade Barbed Reducing Straight Connectors
Use these fittings for a wide range of applications. Ark Plas Products™ Natural Polypropylene Commercial Grade Barbed Reducing Straight Connectors are designed as a low cost alternative with high end performance.
Brand: Ark Plas Products™ CAD34-PR0
Additional Details : Weight : 0.04500kg
Description
- Economic and reliable
- Up to 125psi without clamp
- Contain recycled runner material
- Available in Natural Nylon or Natural Polypropylene
Specifications
Barbed Reducing Straight Connector | |
Polypropylene | |
3/8 x 1/8 in. Barb | |
Natural |
Autoclavable | |
25 Pack | |
Straight |