Learn More
Deutsch Neumann™ Rubber Policemen
Provides for quick and easy transfer of residues to glass surfaces. Deutsch Neumann™ Rubber Policemen are manufactured from natural rubber.
9.05 SEK
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10028230
|
Deutsch Neumann™
2250001 |
Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- 5mm hole for insert
- Available in grey or red color
- Durometer Shore A 40
- Excellent tensile strength and resilience
Specifications
Rubber |