missing translation for 'onlineSavingsMsg'
Learn More

NCAM1/CD56 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E8C9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60238-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887265

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins.
Specifications

Specifications

NCAM1/CD56
Monoclonal
0.95 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13591, P13595, P13596
Ncam1
NCAM1/CD56 Fusion Protein Ag5732
20 μL
Primary
Mouse, Rat, Human, Porcine
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1E8C9
Unconjugated
NCAM1
CD56, MSK39, N CAM 1, NCAM, NCAM 1, NCAM1
Mouse
Protein G
RUO
100515564, 17967, 24586, 4684
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers