missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCKAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12140-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene shares similarity with the mouse Dhm1 and the yeast dhp1 gene. The yeast gene is involved in homologous recombination and RNA metabolism, such as RNA synthesis and RNA trafficking. Complementation studies show that Dhm1 has a similar function in mouse as dhp1. The function of the human gene has not yet been determined. Transcript variants encoding different isoforms have been noted for this gene; however, their full-length nature is not known.Specifications
NCKAP1 | |
Polyclonal | |
Unconjugated | |
NCKAP1 | |
HEM2, KIAA0587, NAP 1, NAP1, NAP125, NCK associated protein 1, NCKAP1, p125Nap1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10787, 50884, 58823 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P28660, P55161, Q9Y2A7 | |
Nckap1 | |
NCKAP1 Fusion Protein Ag2788 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NCKAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction