Learn More
NCLN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10985-1-AP-20UL
 View more versions of this product
Product Code. 16887733
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).
Specifications
Specifications
NCLN
Polyclonal
Unconjugated
NCLN
NCLN, Nicalin, nicalin homolog (zebrafish), Nicastrin like protein
Rabbit
Antigen Affinity Chromatography
RUO
103425, 314648, 56926
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5XIA1, Q8VCM8, Q969V3
Ncln
NCLN Fusion Protein Ag1430
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NCLN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated