Learn More
NDC80 Mouse anti-Human, Unconjugated, Clone: 2B7A8, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NDC80 |
---|---|
Clone | 2B7A8 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889285
|
Proteintech
66960-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879285
|
Proteintech
66960-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NDC80 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NDC80. This antibody recognizes human, rat, mouse antigen. The NDC80 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NDC80 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
10403 | |
Primary |
2B7A8 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.