missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDC80 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18932-1-AP-150UL
This item is not returnable.
View return policy
Description
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.Specifications
NDC80 | |
Polyclonal | |
Unconjugated | |
NDC80 | |
HEC, HEC1, HsHec1, hsNDC80, Kinetochore protein Hec1, KNTC2, NDC80, TID3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10403, 301701, 67052 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14777, Q9D0F1 | |
Ndc80 | |
NDC80 Fusion Protein Ag8065 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NDC80 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction