Learn More
NDUFC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23842-1-AP-20UL
 View more versions of this product
Product Code. 16822475
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
Specifications
NDUFC1
Polyclonal
Unconjugated
NDUFC1
CI KFYI, Complex I KFYI, KFYI, NDUFC1
Rabbit
Antigen Affinity Chromatography
RUO
4717
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43677
NDUFC1
NDUFC1 Fusion Protein Ag20835
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NDUFC1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated