missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15224-1-AP-150UL
This item is not returnable.
View return policy
Description
Proteintech's NDUFS5 antibody 15224-1-AP is capable of detecting the 15 kDa protein in Western blotting.
Located in the mitochondrial inner membrane, mitochondrial complex I is the first and largest enzyme in the electron transport chain of oxidative phosphorylation. By oxidizing NADH that is produced in the Krebs cycle, this complex utilizes the two electrons to reduce ubiquinone to ubiquinol, thereby initiating the passage of electrons to successive complexes and ultimately leading to the reduction of oxygen to water. Mitochondrial complex I consists of over 40 subunits and is of considerable clinical interest since defects in any of the subunits can lead to various myopathies and neuropathies. As a subunit of mitochondrial complex I, NDUFS5 (NADH dehydrogenase [ubiquinone] iron-sulfur protein 5), also designated NADH-ubiquinone oxidoreductase 15 kDa subunit, is a 106 amino acid protein that is suggested to not be involved in catalysis. NDUFS5 is expressed ubiquitously in human tissues, with a relative higher expression in human heart, skeletal muscle, liver, kidney and fetal heart.Specifications
NDUFS5 | |
Polyclonal | |
Unconjugated | |
NDUFS5 | |
CI 15 kDa, Complex I 15 kDa, NDUFS5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362588, 4725, 595136 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43920, Q99LY9 | |
Ndufs5 | |
NDUFS5 Fusion Protein Ag7441 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NDUFS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction