missing translation for 'onlineSavingsMsg'
Learn More

NEK9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen NEK9
Concentration 0.25 mg/mL
Applications Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888103
View Documents
Proteintech
11192-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16878103
View Documents
Proteintech
11192-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NEK9, a NEK type protein kinase, regulates chromosome alignment and segregation in mitosis. The protein has a N-terminal NIMA-like catalytic domain, a central domain with homology to the guanine nucleotide exchange factor for the GTPase Ran (RCC1), and a C-terminal coiled-coil domain. It is phosphorylated by active p34(Cdc2) and is capable of autophosphorylation and oligomerization. The activated protein binds Nek6 and RCC1, and has been shown to associate with the human homolog of the Drosophila protein Bicaudal D (Bicd2) in vivo and to phosphorylate this coiled-coil protein in vitro. NEK9 expression has been documented in normal human brain. ESTs have been isolated from several tissue libraries, including normal bone, lymph and thymus and cancerous cervix, eye, lymph and placenta.
Specifications

Specifications

NEK9
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
Unconjugated
Rabbit
Human, Rat, Mouse
Q8K1R7, Q8TD19
217718, 299204, 91754
NEK9 Fusion Protein Ag1678
Primary
-20°C
NEK9
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
KIAA1995, Nek8, NEK9, NERCC, NERCC1, Nercc1 kinase, NimA related kinase 8, NimA related protein kinase 9
NEK9
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NEK9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.