Learn More
NEU3 Mouse anti-Human, Unconjugated, Clone: 2C9C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67098-1-IG-150UL
This item is not returnable.
View return policy
Description
The NEU3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEU3. This antibody recognizes human antigen. The NEU3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
NEU3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
2C9C5 | |
Unconjugated | |
150 μL | |
10825 | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NEU3 Mouse anti-Human, Unconjugated, Clone: 2C9C5, Proteintech > 150 μL
Spot an opportunity for improvement?Share a Content Correction