missing translation for 'onlineSavingsMsg'
Learn More

NEUROD1 Mouse anti-Human, Clone: 3E10F1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66691-1-IG-150UL

 View more versions of this product

Product Code. 16878805

  • 4652.90 SEK / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NeuroD1 encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus.
Specifications

Specifications

NEUROD1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13562
NEUROD1
NEUROD1 Fusion Protein Ag27606
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
3E10F1
Unconjugated
NEUROD1
BETA2, BHF 1, bHLHa3, NEUROD, NEUROD1, neurogenic differentiation 1
Mouse
Protein G
RUO
4760
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NEUROD1 Mouse anti-Human, Clone: 3E10F1, Proteintech > 150 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.