Learn More
Neuromedin B Mouse anti-Human, Mouse, Clone: 1G3B6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66781-1-IG-150UL
This item is not returnable.
View return policy
Description
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.Specifications
Neuromedin B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08949, Q9CR53 | |
NMB | |
Neuromedin B Fusion Protein Ag0842 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
1G3B6 | |
Unconjugated | |
NMB | |
neuromedin B, NMB | |
Mouse | |
Protein G | |
RUO | |
4828, 68039 | |
-20°C | |
Liquid |
Product Content Correction
Product Title
Neuromedin B Mouse anti-Human, Mouse, Clone: 1G3B6, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction