missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroserpin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12558-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. The protein is primarily secreted by axons in the brain, and preferentially reacts with and inhibits tissue-type plasminogen activator. It is thought to play a role in the regulation of axonal growth and the development of synaptic plasticity. Mutations in this gene result in familial encephalopathy with neuroserpin inclusion bodies (FENIB), which is a domitly inherited form of familial encephalopathy and epilepsy characterized by the accumulation of mutant neuroserpin polymers. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
Neuroserpin | |
Polyclonal | |
Unconjugated | |
SERPINI1 | |
neuroserpin, Peptidase inhibitor 12, PI 12, PI12, Serpin I1, SERPINI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116459, 20713, 5274 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35684, Q99574, Q9JLD2 | |
SERPINI1 | |
Neuroserpin Fusion Protein Ag3275 | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Neuroserpin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction