Learn More
NeutraKine™ IFN Alpha 2A Mouse anti-Human, Unconjugated, Clone: 2H8G1, Proteintech
Mouse Monoclonal Antibody
1690.93 SEK - 3620.19 SEK
Specifications
Antigen | NeutraKine™ IFN Alpha 2A |
---|---|
Clone | 2H8G1 |
Applications | Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800426
|
Proteintech
69008-1-IG-25UG |
25 ÎĽg |
1690.93 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890416
|
Proteintech
69008-1-IG-100UG |
100 ÎĽg |
3620.19 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™ IFN Alpha 2A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IFN Alpha 2A. This antibody recognizes human antigen. The NeutraKine™ IFN Alpha 2A antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
NeutraKine™ IFN Alpha 2A | |
Neutralization | |
Unconjugated | |
Human | |
IgG1 |
2H8G1 | |
Monoclonal | |
Mouse | |
3440 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.