Learn More
NeutraKine™ IL-4 Mouse anti-Human, Unconjugated, Clone: 1H5G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69005-1-IG-100UG
Additional Details : Weight : 0.00010kg
Description
The NeutraKine™ IL-4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-4. This antibody recognizes human antigen. The NeutraKine™ IL-4 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
NeutraKine™ IL-4 | |
Monoclonal | |
Unconjugated | |
100 μg | |
3565 | |
IgG1 |
Neutralization | |
1H5G5 | |
Mouse | |
Primary | |
Human |