Learn More
NeutraKine™ IL-4 Mouse anti-Human, Unconjugated, Clone: 1H5G5, Proteintech
Mouse Monoclonal Antibody
1690.93 SEK - 3620.19 SEK
Specifications
Antigen | NeutraKine™ IL-4 |
---|---|
Clone | 1H5G5 |
Applications | Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840416
|
Proteintech
69005-1-IG-25UG |
25 ÎĽg |
1690.93 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16830416
|
Proteintech
69005-1-IG-100UG |
100 ÎĽg |
3620.19 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™ IL-4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-4. This antibody recognizes human antigen. The NeutraKine™ IL-4 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
NeutraKine™ IL-4 | |
Neutralization | |
Unconjugated | |
Human | |
IgG1 |
1H5G5 | |
Monoclonal | |
Mouse | |
3565 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.