Learn More
Macherey-Nagel™ Neutralization Buffer PFN
Neutralization buffer to be used with the NucleoSpin VET in combination with NucleoProtect VET products or with the NucleoSpin RNA Plant and Fungi kit.
Brand: Macherey-Nagel™ 740121.5
Alert:
rna, purification, isolation, nucleospin, plant, fungi, fruit, bead, tube, acidic
Description
- The recommended volume of buffer PFN depends on the sample type:
- Kiwi: 50 ÎĽL
- Lemon: 50 ÎĽL
- Apple: 15 ÎĽL
- Orange: 15 ÎĽL
- Blueberry: 50 ÎĽL
- Fungal hyphae: 0–50 μL
- During storage, especially at low temperatures, a precipitate may form, which can be easily dissolved by incubating the bottle at 40 °C before use
Specifications
Colorless | |
tris(hydroxymethyl)aminomethane and water | |
77-86-1 | |
Solution |
NucleoSpin RNA Plant and Fungi Kit | |
5 mL | |
Odorless | |
Processing acidic samples for RNA isolation |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.