Learn More
Eppendorf™ Scientific Shaker Test Tube Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Brand: Eppendorf™ M1289-0100
3551.32 SEK valid until 2024-12-31
Use promo code "21970" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
8 to 11 mm | |
Stainless Steel | |
48 |
Test Tube Rack | |
Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series | |
Large |