Learn More
NFKBIE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11273-1-AP-20UL
 View more versions of this product
Product Code. 16888233
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. Inhibits DNA-binding of NF-kappa-B p50-p65 and p50-c-Rel complexes.
Specifications
Specifications
NFKBIE
Polyclonal
Unconjugated
NFKBIE
I kappa B epsilon, IkappaBepsilon, IkB E, IkB epsilon, IKBE, NF kappa B inhibitor epsilon, NF kappa BIE, NFKBIE
Rabbit
Antigen Affinity Chromatography
RUO
18037, 316241, 4794
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00221, O54910
Nfkbie
NFKBIE Fusion Protein Ag1800
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NFKBIE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated