missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
NG2/MCSP Antibody (LHM 2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB100-2688SS
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NG2/MCSP Monoclonal specifically detects NG2/MCSP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated.
Specifikationer
| NG2/MCSP | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Flow (Cell Surface) 1 - 2.5 ug/mL | |
| chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Other species have not been tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| LHM 2 | |
| Unconjugated | |
| Q6UVK1 | |
| CSPG4 | |
| A 375P cells crude extract | |
| 0.025 mL | |
| Cancer, Neuroscience | |
| 1464 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering