missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR/TNFRSF16/p75NTR Rabbit anti-Human, Clone: NGFR/2550R, Novus Biologicals™
Rabbit Monoclonal Antibody
2815.00 SEK
Specifications
Antigen | NGFR/TNFRSF16/p75NTR |
---|---|
Clone | NGFR/2550R |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
NGFR/TNFRSF16/p75NTR | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
4804 | |
IgG | |
Protein A or G purified |
NGFR/2550R | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
Recombinant human NGFR/TNFRSF16/p75NTR protein fragment (around aa 281-421) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NGFR/TNFRSF16/p75NTR Rabbit anti-Human, Clone: NGFR/2550R, Novus Biologicals™