missing translation for 'onlineSavingsMsg'
Learn More
β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate Product Code.: 15815198

β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate

Product Code. 15815198 Shop All Thermo Scientific Products
1 g, Plastic bottle, 1g
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Plastic bottle
Unit Size:
1g
250mg
500mg
5g
This item is not returnable. View return policy

Product Code. 15815198

Brand: Thermo Scientific J22655.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1184-16-3 | C21H27N7NaO17P3 | 765.39 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-16-3
Molecular Formula C21H27N7NaO17P3
Molecular Weight (g/mol) 765.39
MDL Number MFCD00036973
InChI Key JNUMDLCHLVUHFS-QYZPTAICSA-M
Synonym nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
PubChem CID 131845262
IUPAC Name [[(2 R,3 R,4 R,5 R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2 R,3 S,4 R,5 R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodiÎĽm
SMILES [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Physical Form Powder
Quantity 1 g
Beilstein 4779954
Formula Weight 837.45
Packaging Plastic bottle
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt tetrahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number190735 Certificate TypeCertificate of Analysis Date27/11/2022 Product CodeJ22655.03,J22655.ME,J22655.MD
Lot Number185824 Certificate TypeCertificate of Analysis Date27/11/2022 Product CodeJ22655.03,J22655.ME,J22655.MD
Lot Number181141 Certificate TypeCertificate of Analysis Date27/11/2022 Product CodeJ22655.03,J22655.ME,J22655.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.