Läs mer
Niflumic acid, 99+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60489.18
| Kvantitet | 50g |
|---|
Se fler versioner av denna produkt
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.
Solubility
Soluble in ethanol (∼50 mg/ml), acetone (50 mg/ml, Clear to Slightly Hazy, Yellow), methanol (∼50 mg/ml), DMSO (56 mg/ml at 25°C), and acetonitrile (∼50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Hygroscopic. Keep away fom strong oxidizing agents.
Kemiska identifierare
| 4394-00-7 | |
| 282.22 | |
| JZFPYUNJRRFVQU-UHFFFAOYSA-N | |
| 4488 | |
| 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid |
| C13H9F3N2O2 | |
| MFCD00010569 | |
| niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french | |
| CHEBI:34888 | |
| OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F |
Specifikationer
| Niflumic acid | |
| C13H9F3N2O2 | |
| 50g | |
| Light sensitive | |
| niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french | |
| JZFPYUNJRRFVQU-UHFFFAOYSA-N | |
| 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid | |
| 4488 | |
| 282.22 |
| 4394-00-7 | |
| MFCD00010569 | |
| UN2811 | |
| 14,6531 | |
| Soluble in ethanol (∽50mg/ml),acetone (50mg/ml,Clear to Slightly Hazy,Yellow),methanol (∽50mg/ml),DMSO (56mg/ml at 25°C),and acetonitrile (∽50mg/ml). | |
| OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F | |
| 282.22 | |
| CHEBI:34888 | |
| ≥99% |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.