Learn More
NIPBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18792-1-AP-20UL
 View more versions of this product
Product Code. 16898624
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
Specifications
Specifications
NIPBL
Polyclonal
Unconjugated
NIPBL
CDLS, CDLS1, Delangin, IDN3, IDN3 B, NIPBL, Nipped B homolog (Drosophila), Nipped B like protein, SCC2 homolog
Rabbit
Antigen Affinity Chromatography
RUO
25836, 294787, 71175
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6KC79, Q6KCD5
Nipbl
NIPBL Fusion Protein Ag5663
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NIPBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated