Learn More
NIPBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18792-1-AP-20UL
This item is not returnable.
View return policy
Description
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.Specifications
NIPBL | |
Polyclonal | |
Unconjugated | |
NIPBL | |
CDLS, CDLS1, Delangin, IDN3, IDN3 B, NIPBL, Nipped B homolog (Drosophila), Nipped B like protein, SCC2 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25836, 294787, 71175 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6KC79, Q6KCD5 | |
Nipbl | |
NIPBL Fusion Protein Ag5663 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
NIPBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction