missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13013-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.Specifications
NKX2-2 | |
Polyclonal | |
Unconjugated | |
NKX2-2 | |
Homeobox protein Nkx 2.2, NK2 homeobox 2, NKX2 2, NKX2.2, NKX2-2, NKX2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18088, 366214, 4821 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95096, P42586 | |
Nkx2-2 | |
NKX2-2 Fusion Protein Ag4069 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NKX2-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction