missing translation for 'onlineSavingsMsg'
Learn More

NKX2-2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13013-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881014

  • 1656.73 SEK / 20µL
Estimated Shipment: 19-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Specifications

Specifications

NKX2-2
Polyclonal
Unconjugated
NKX2-2
Homeobox protein Nkx 2.2, NK2 homeobox 2, NKX2 2, NKX2.2, NKX2-2, NKX2B
Rabbit
Antigen Affinity Chromatography
RUO
18088, 366214, 4821
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95096, P42586
Nkx2-2
NKX2-2 Fusion Protein Ag4069
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers