Learn More
NMNAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13236-1-AP-20UL
 View more versions of this product
Product Code. 16881354
  • 1645.54 SEK / 20µL
Description
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate with the same efficiency. Can use triazofurin monophosphate (TrMP) as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity, can use NAD (+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+). Protects against axonal degeneration following injury.
Specifications
Specifications
NMNAT3
Polyclonal
Unconjugated
NMNAT3
NaMN adenylyltransferase 3, NMNAT3, PNAT 3, PNAT3
Rabbit
Antigen Affinity Chromatography
RUO
349565, 363118, 74080
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96T66, Q99JR6
NMNAT3
NMNAT3 Fusion Protein Ag3965
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
NMNAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated