missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMU Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24862-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
may induce uterine smooth muscle contraction and selective vasoconstriction.Specifications
NMU | |
Polyclonal | |
Unconjugated | |
NMU | |
neuromedin U, Neuromedin-U-25, NMU | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10874 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48645 | |
NMU | |
NMU Fusion Protein Ag21516 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |