missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMU Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NMU |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883355
|
Proteintech
24862-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16873355
|
Proteintech
24862-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
may induce uterine smooth muscle contraction and selective vasoconstriction.Specifications
NMU | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P48645 | |
10874 | |
NMU Fusion Protein Ag21516 | |
Primary | |
-20°C | |
NMU |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
neuromedin U, Neuromedin-U-25, NMU | |
NMU | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |