Learn More
Non-Return Valve for Vacuum Lines
Designed for use in vacuum lines and fluid handling systems
Brand:  Azlon NWH902P
 View more versions of this product
Product Code. 10208114
  • 533.00 SEK / Pack of 5
Only null left
Estimated Shipment: 09-09-2025
Description
  • Neoprene disc valve has no spring and will operate at very low back pressures (NWH905P valve is now manufactured from nitrile rubber)
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing
  • Easily disassembled for cleaning
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 to 7 mm
Polypropylene, Neoprene
For preventing back pressure in filter flasks
Non-return Valve
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Non-Return Valve for Vacuum Lines > Diameter: 4 to 7mm