Learn More
NONO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11058-1-AP-20UL
 View more versions of this product
Product Code. 16887873
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes an RNA-binding protein which plays various roles in the nucleus, including transcriptional regulation and RNA splicing. A rearrangement between this gene and the transcription factor E3 gene has been observed in papillary renal cell carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes exist on Chromosomes 2 and 16.
Specifications
Specifications
NONO
Polyclonal
Unconjugated
NONO
55 kDa nuclear protein, NONO, NonO protein, NRB54, P54, p54(nrb), P54NRB
Rabbit
Antigen Affinity Chromatography
RUO
317259, 4841, 53610
-20°C
Liquid
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15233, Q5FVM4, Q99K48
Nono
NONO Fusion Protein Ag1512
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NONO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated