missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
NOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17772-1-AP-150UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.Specifikationer
| NOX1 | |
| Polyclonal | |
| Unconjugated | |
| NOX1 | |
| GP91 2, Mitogenic oxidase 1, MOX 1, MOX1, NADPH oxidase 1, NOH 1, NOH1, NOX 1, NOX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114243, 237038, 27035 | |
| -20°C | |
| Liquid |
| ELISA, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CIZ9, Q9WV87, Q9Y5S8 | |
| Nox1 | |
| NOX1 Fusion Protein Ag12187 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering