missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11960-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.Specifications
NPM3 | |
Polyclonal | |
Unconjugated | |
NPM3 | |
NPM3, nucleophosmin/nucleoplasmin, 3, Nucleoplasmin 3, PORMIN, TMEM123 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10360, 18150, 502389 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75607, Q9CPP0 | |
Npm3 | |
NPM3 Fusion Protein Ag2564 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NPM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction