missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NPT1 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890315
|
Proteintech
20751-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880315
|
Proteintech
20751-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Western blot analysis using this antibody detected a major band around 45-55 kDa in kidney tissue.
The function of this protein remains unknown.Specifications
NPT1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q14916, Q61983, Q62795 | |
171080, 20504, 6568 | |
NPT1 Fusion Protein Ag12819 | |
Primary | |
-20°C | |
SLC17A1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Na(+)/PI cotransporter 1, Na/Pi 4, NAPI 1, SLC17A1 | |
Slc17a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech