missing translation for 'onlineSavingsMsg'
Learn More
NSC 23766 trihydrochloride, MedChemExpress Product Code.: 16496768

NSC 23766 trihydrochloride, MedChemExpress

Product Code. p-200097008
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496768

Brand: Medchem Express HY15723A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC 23766 trihydrochloride is an inhibitor of Rac1 activation.

NSC 23766 trihydrochloride is an inhibitor of Rac1 activation.

Specifications

Chemical Name or Material NSC 23766 trihydrochloride
CAS 1177865-17-6
Color White
Purity Grade Notes Research
Molecular Formula C24H38Cl3N7
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (62.77 mM; Need ultrasonic) ∣H2O : ≥ 32 mg/mL (60.27 mM)
SMILES NC1=CC(C)=NC2=CC=C(NC3=NC(NC(C)CCCN(CC)CC)=NC(C)=C3)C=C12.[H]Cl.[H]Cl.[H]Cl
Molecular Weight (g/mol) 530.96
Formula Weight 530.96
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
NSC 23766 trihydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.