missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5DC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NT5DC3 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884755
|
Proteintech
26242-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874755
|
Proteintech
26242-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NT5DC3 gene ontology annotations related to this gene include 5′-nucleotidase activity; metal ion binding.Specifications
NT5DC3 | |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q86UY8 | |
51559 | |
NT5DC3 Fusion Protein Ag23962 | |
Primary | |
-20°C | |
NT5DC3 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GNN, GRP94 neighboring nucleotidase, NT5DC3, TU12B1 TY, TU12B1TY | |
NT5DC3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NT5DC3 Rabbit anti-Human, Polyclonal, Proteintech