missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUCB2/nesfatin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26712-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a leucine-rich cytoplasmic protein, which is highly similar to a mouse protein that negatively regulates Ca/calmodulin-dependent protein kinase II phosphorylation and may be essential for spatial learning processes.Specifications
NUCB2/nesfatin-1 | |
Polyclonal | |
Unconjugated | |
NUCB2 | |
DNA binding protein NEFA, Gastric cancer antigen Zg4, NEFA, NUCB2, nucleobindin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4925, 53322, 59295 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P80303, P81117, Q9JI85 | |
Nucb2 | |
NUCB2/nesfatin-1 Fusion Protein Ag25152 | |
20 ÎĽL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NUCB2/nesfatin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction