missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUCKS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12023-2-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a nuclear protein that is highly conserved in vertebrates. The conserved regions of the protein contain several consensus phosphorylation sites for casein kinase II and cyclin-dependent kinases, two putative nuclear localization signals, and a basic DNA-binding domain. It is phosphorylated in vivo by Cdk1 during mitosis of the cell cycle.Specifications
NUCKS1 | |
Polyclonal | |
Unconjugated | |
NUCKS1 | |
JC7, NUCKS, NUCKS1, P1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64709, 64710, 98415 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80XU3, Q9H1E3 | |
Nucks1 | |
NUCKS1 Fusion Protein Ag2652 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NUCKS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction